Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001113355.1 | NP_001106826.1 | 4 | 574 | |
NM_133672.3 | NP_598433.1 | 4 | 501 | ||
GenBank mRNA | AK028096.1 | - | 4 | 575 | |
AK086851.1 | - | 397 | |||
AK147989.1 | - | 4 | 442 | ||
AK149859.1 | - | 4 | 426 | ||
AK150244.1 | - | 4 | 399 | ||
AK151360.1 | - | 4 | 411 | ||
AK152424.1 | - | 4 | 445 | ||
AK159783.1 | - | 4 | 451 | ||
AK165853.1 | - | 4 | 445 | ||
AK167592.1 | - | 4 | 456 | ||
AK168213.1 | - | 4 | 501 | ||
AK168537.1 | - | 4 | 456 | ||
AK216993.1 | - | 3 | 364 | ||
BC007148.1 | - | 4 | 422 | 3485844 | |
BC058236.1 | - | 3 | 352 | ||
S41204.1 | - | 4 | 448 |