Gene Transcripts

Gene Symbol
Zmat3
Entrez Gene ID
22401
Gene Name
zinc finger matrin type 3
Gene Aliases
Wig1
Chromosome Location
Chr. 3: 32334792 - 32366008 on Build GRCm38
UniGene ID
Mm.35705
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009517.2 NP_033543.2 6 1721
GenBank mRNA AF012923.1 - 6 1673
BC138740.1 - 5 1540 8861762
BC138742.1 - 5 1540 8861764

View Full Product Details