Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_009531.2 | NP_033557.2 | 7 | 963 | |
GenBank mRNA | AB071144.1 | - | 7 | 900 | |
AK004713.1 | - | 7 | 903 | ||
AK028595.1 | - | 7 | 901 | ||
AK166981.1 | - | 7 | 898 | ||
AK170975.1 | - | 7 | 895 | ||
BC052725.1 | - | 7 | 925 | 6812198 | |
U27398.1 | - | 7 | 871 | ||
U40005.1 | - | 6 | 702 |