Gene Transcripts

Gene Symbol
Zfand5
Entrez Gene ID
22682
Gene Name
zinc finger, AN1-type domain 5
Gene Aliases
2310057A04Rik, 5830475F03Rik, AA415485, Za20d2, Zfp216
Chromosome Location
Chr. 19: 21271279 - 21286840 on Build GRCm38
UniGene ID
Mm.292405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009551.5 NP_033577.1 6 1573
GenBank mRNA AF062071.1 - 5 1165
AK076397.1 - 6 1574
AK131911.1 - 6 1569
BC080270.1 - 6 1554

View Full Product Details