Gene Transcripts

Gene Symbol
Tbx5
Entrez Gene ID
21388
Gene Name
T-box 5
Gene Aliases
Chromosome Location
Chr. 5: 119796515 - 119885220 on Build GRCm38
UniGene ID
Mm.103636
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011537.3 NP_035667.1 8, 9 1638
GenBank mRNA AF140427.1 - 8, 9 1403
AK044843.1 - 4, 5 617
BC090639.1 - 8, 9 1642 30548234

View Full Product Details