Gene Transcripts

Gene Symbol
Tsks
Entrez Gene ID
22116
Gene Name
testis-specific serine kinase substrate
Gene Aliases
Stk22s1, Tssks1
Chromosome Location
Chr. 7: 44929577 - 44958035 on Build GRCm38
UniGene ID
Mm.84502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001077591.1 NP_001071059.1 8 1312
NM_011651.2 NP_035781.2 8 1339
GenBank mRNA AF025310.2 - 8 1319
BC118618.1 - 8 1291 40107271
BC121827.1 - 8 1291 40107270

View Full Product Details