Gene Transcripts

Gene Symbol
Ugcg
Entrez Gene ID
22234
Gene Name
UDP-glucose ceramide glucosyltransferase
Gene Aliases
AU043821, C80537, Epcs21, GlcT-1, Ugcgl
Chromosome Location
Chr. 4: 59189204 - 59222833 on Build GRCm38
UniGene ID
Mm.198803
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011673.3 NP_035803.1 2 297
GenBank mRNA AK138848.1 - 2 313
BC050828.1 - 2 297 6512920
D89866.1 - 2 388

View Full Product Details