Gene Transcripts
- Gene Symbol
- Vmn1r51
- Entrez Gene ID
- 22296
- Gene Name
- vomeronasal 1 receptor 51
- Gene Aliases
- V1R, V1r1, V1ra1, VN12, mV1R1
- Chromosome Location
- Chr. 6: 90122643 - 90130645 on Build GRCm38
- UniGene ID
- Mm.20471
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_011683.2 |
NP_035813.2
|
3 |
2333 |
|
GenBank mRNA |
Y12725.1 |
- |
2 |
2303 |
|
View Full Product Details