Gene Transcripts

Gene Symbol
Lin7b
Entrez Gene ID
22342
Gene Name
lin-7 homolog B (C. elegans)
Gene Aliases
LIN-7B, MALS-2, Veli2
Chromosome Location
Chr. 7: 45367891 - 45370564 on Build GRCm38
UniGene ID
Mm.20472
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_011698.1 NP_035828.1 4 360
GenBank mRNA AF087694.1 - 4 367
AF173082.1 - 4 360
AK019299.1 - 4 365
BC031780.1 - 4 379 4164883

View Full Product Details