Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_012001.2 | NP_036131.1 | 2 | 273 | |
GenBank mRNA | AF071314.1 | - | 2 | 142 | |
AK003752.1 | - | 2 | 135 | ||
AK009955.1 | - | 2 | 145 | ||
AK162763.1 | - | 2 | 139 | ||
AK167833.1 | - | 2 | 159 | ||
BC116800.1 | - | 2 | 127 | 40126119 | |
BC116826.1 | - | 2 | 127 | 40126145 | |
CX203220.1 | - | 2 | 273 |