Gene Transcripts

Gene Symbol
NUDT21
Entrez Gene ID
11051
Gene Name
nudix hydrolase 21
Gene Aliases
CFIM25, CPSF5
Chromosome Location
Chr. 16: 56429136 - 56451349 on Build GRCh38
UniGene ID
Hs.528834
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007006.2 NP_008937.1 3 495
GenBank mRNA AJ001810.1 - 3 348
AK312721.1 - 3 498
AL535307.3 - 3 495
BC001403.2 - 3 456 3139379
BX537360.1 - 3 499
CR456919.1 - 3 348

View Full Product Details