Gene Transcripts

Gene Symbol
Azi2
Entrez Gene ID
27215
Gene Name
5-azacytidine induced gene 2
Gene Aliases
AA410145, AZ2
Chromosome Location
Chr. 9: 118040401 - 118069798 on Build GRCm38
UniGene ID
Mm.92705
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048146.2 NP_001041611.1 3 747
NM_001286507.1 NP_001273436.1 3 747
NM_001286508.1 NP_001273437.1 3 665
NM_013727.4 NP_038755.1 3 747
GenBank mRNA AB007141.1 - 3 583
AK015942.1 - 3 568
AK030076.1 - 3 579
AK036102.1 - 3 654
AK134714.1 - 3 650
AK153477.1 - 3 711
AK165549.1 - 3 633
AK167783.1 - 3 691
BC006851.1 - 3 642 3156857

View Full Product Details