Gene Transcripts

Gene Symbol
Rps26
Entrez Gene ID
27370
Gene Name
ribosomal protein S26
Gene Aliases
Chromosome Location
Chr. 10: 128624529 - 128626506 on Build GRCm38
UniGene ID
Mm.261679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013765.2 NP_038793.2 4 375
GenBank mRNA AK008301.1 - 4 371
AK010689.2 - 4 370
AK012722.1 - 4 373
AK018702.1 - 4 371
AK152132.1 - 4 371
AV585706.1 - 3 329
BC036987.1 - 3 361 4987659
BC071192.1 - 1215
BC081452.1 - 3 348 6411109
BC100456.1 - 4 362 6541446
BU530648.1 - 4 376
U67770.1 - 3 352

View Full Product Details