Gene Transcripts

Gene Symbol
Mmel1
Entrez Gene ID
27390
Gene Name
membrane metallo-endopeptidase-like 1
Gene Aliases
Mell1, NEP2, NEPII, Nl1, SEP
Chromosome Location
Chr. 4: 154869585 - 154895530 on Build GRCm38
UniGene ID
Mm.116944
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013783.2 NP_038811.2 3, 4 529
GenBank mRNA AF157105.1 - 3, 4 475
AF176569.1 - 3, 4 526
AF302076.1 - 2, 3 219

View Full Product Details