Gene Transcripts

Gene Symbol
Trpc3
Entrez Gene ID
22065
Gene Name
transient receptor potential cation channel, subfamily C, member 3
Gene Aliases
Mwk, Trcp3, Trp3, Trrp3
Chromosome Location
Chr. 3: 36620482 - 36690229 on Build GRCm38
UniGene ID
Mm.74363
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019510.2 NP_062383.2 5 1686
GenBank mRNA AF190645.1 - 4 1276
AK032317.1 - 5 1687
AK080619.1 - 5 1674

View Full Product Details