Gene Transcripts
- Gene Symbol
- Tcte2
- Entrez Gene ID
- 21646
- Gene Name
- t-complex-associated testis expressed 2
- Gene Aliases
- AI429268, Tcte-2
- Chromosome Location
- Chr. 17: 13716436 - 13761394 on Build GRCm38
- UniGene ID
- Mm.486358
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK008572.1 |
- |
|
509 |
|
View Full Product Details