Gene Transcripts

Gene Symbol
Upk3a
Entrez Gene ID
22270
Gene Name
uroplakin 3A
Gene Aliases
1110017C07Rik, UP3a, UPIII, Upk3
Chromosome Location
Chr. 15: 85017141 - 85022560 on Build GRCm38
UniGene ID
Mm.46214
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023478.2 NP_075967.2 2 254
GenBank mRNA AF222750.2 - 2 175
BC125336.1 - 2 254 40129851
BC125338.1 - 2 254 40129853
BX512009.1 - 1 61
DQ387454.1 - 2 204

View Full Product Details