Gene Transcripts

Gene Symbol
Cyp2c7
Entrez Gene ID
29298
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 7
Gene Aliases
Cyp2c39
Chromosome Location
Chr. 1: 147422999 - 148119979 on Build Rnor_6.0
UniGene ID
Rn.1247
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017158.2 NP_058854.1 3 402
GenBank mRNA M18335.1 - 3 403
M31031.1 - 3 360

View Full Product Details