Gene Transcripts

Gene Symbol
Rab4b
Entrez Gene ID
50866
Gene Name
RAB4B, member RAS oncogene family
Gene Aliases
Chromosome Location
Chr. 1: 83979298 - 83990588 on Build Rnor_6.0
UniGene ID
Rn.7181
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017355.1 NP_059051.1 6 544
XM_008759181.1 XP_008757403.1 5 763
XM_008759182.1 XP_008757404.1 5 763
GenBank mRNA FQ227674.1 - 6 576
X78605.1 - 6 544

View Full Product Details