Gene Transcripts
- Gene Symbol
- LILRA2
- Entrez Gene ID
- 11027
- Gene Name
- leukocyte immunoglobulin like receptor A2
- Gene Aliases
- CD85H, ILT1, LIR-7, LIR7
- Chromosome Location
- Chr. 19: 54572997 - 54587560 on Build GRCh38
- UniGene ID
- Hs.655593
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U82275.1 |
- |
7 |
1507 |
|
U82276.1 |
- |
6 |
1422 |
|
U82277.1 |
- |
5 |
631 |
|
U82278.1 |
- |
8 |
1471 |
|
View Full Product Details