Gene Transcripts

Gene Symbol
Cybb
Entrez Gene ID
66021
Gene Name
cytochrome b-245, beta polypeptide
Gene Aliases
Gp91-phox, Nox2
Chromosome Location
Chr. X: 14578330 - 14610049 on Build Rnor_6.0
UniGene ID
Rn.98491
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023965.1 NP_076455.1 4 271
GenBank mRNA AF298656.3 - 4 271
AJ295950.1 - 4 271
FQ217794.1 - 340
FQ231233.1 - 324

View Full Product Details