Gene Transcripts
- Gene Symbol
- Csf1
- Entrez Gene ID
- 78965
- Gene Name
- colony stimulating factor 1 (macrophage)
- Gene Aliases
- Chromosome Location
- Chr. 2: 210522370 - 210550546 on Build Rnor_6.0
- UniGene ID
- Rn.83632
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_023981.4 |
NP_076471.3
|
2, 3 |
321 |
|
XM_006233187.1 |
XP_006233249.1
|
2, 3 |
192 |
|
XM_008761426.1 |
XP_008759648.1
|
2, 3 |
192 |
|
XM_008761427.1 |
XP_008759649.1
|
2, 3 |
192 |
|
XM_008761428.1 |
XP_008759650.1
|
2, 3 |
192 |
|
XM_008761429.1 |
XP_008759651.1
|
2, 3 |
192 |
|
GenBank mRNA |
AF514355.1 |
- |
2, 3 |
161 |
|
AF514356.1 |
- |
2, 3 |
161 |
|
AF514357.1 |
- |
2, 3 |
171 |
|
AF515736.1 |
- |
2, 3 |
259 |
|
BC074007.1 |
- |
2, 3 |
321 |
7099454 |
View Full Product Details