Gene Transcripts

Gene Symbol
Akap4
Entrez Gene ID
79254
Gene Name
A kinase (PRKA) anchor protein 4
Gene Aliases
Chromosome Location
Chr. X: 16296027 - 16306078 on Build Rnor_6.0
UniGene ID
Rn.10777
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024402.1 NP_077378.1 4 308
GenBank mRNA AF008114.1 - 4 308

View Full Product Details