Gene Transcripts
- Gene Symbol
- BTN2A2
- Entrez Gene ID
- 10385
- Gene Name
- butyrophilin subfamily 2 member A2
- Gene Aliases
- BT2.2, BTF2, BTN2.2
- Chromosome Location
- Chr. 6: 26382891 - 26394874 on Build GRCh38
- UniGene ID
- Hs.373938
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001197237.1 |
NP_001184166.1
|
5, 6 |
1056 |
|
NM_001197238.1 |
NP_001184167.1
|
5, 6 |
1157 |
|
NM_001197239.1 |
NP_001184168.1
|
3, 4 |
404 |
|
NM_001197240.1 |
NP_001184169.1
|
4, 5 |
752 |
|
NM_006995.4 |
NP_008926.2
|
5, 6 |
1034 |
|
NM_181531.2 |
NP_853509.1
|
4, 5 |
686 |
|
XM_005248797.3 |
XP_005248854.1
|
5, 6 |
1051 |
|
XM_005248798.4 |
XP_005248855.1
|
4, 5 |
1011 |
|
XM_006714953.3 |
XP_006715016.1
|
5, 6 |
1303 |
|
XM_006714955.3 |
XP_006715018.1
|
5, 6 |
1051 |
|
XM_011514228.2 |
XP_011512530.1
|
5, 6 |
1356 |
|
XM_011514229.2 |
XP_011512531.1
|
5, 6 |
865 |
|
XM_011514231.2 |
XP_011512533.1
|
3, 4 |
729 |
|
XM_017010166.1 |
XP_016865655.1
|
5, 6 |
1054 |
|
XM_017010167.1 |
XP_016865656.1
|
4, 5 |
956 |
|
XM_017010168.1 |
XP_016865657.1
|
5, 6 |
1307 |
|
XM_017010169.1 |
XP_016865658.1
|
5, 6 |
1025 |
|
GenBank mRNA |
AK123902.1 |
- |
1, 2 |
101 |
|
AK289883.1 |
- |
4, 5 |
674 |
|
AK293526.1 |
- |
3, 4 |
379 |
|
AK298570.1 |
- |
4, 5 |
727 |
|
BC013258.2 |
- |
3, 4 |
606 |
|
BC014021.2 |
- |
5, 6 |
1032 |
4124138 |
BC017497.1 |
- |
5, 6 |
1049 |
4648935 |
U90550.1 |
- |
5, 6 |
1009 |
|
View Full Product Details