Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031978.1 | NP_114184.1 | 4 | 298 | |
GenBank mRNA | AJ006340.1 | - | 4 | 298 | |
BC087654.1 | - | 4 | 383 | ||
BC105883.1 | - | 4 | 344 | ||
BC128734.1 | - | 4 | 343 | ||
BC166420.1 | - | 4 | 339 |