Gene Transcripts

Gene Symbol
SF3A2
Entrez Gene ID
8175
Gene Name
splicing factor 3a subunit 2
Gene Aliases
PRP11, PRPF11, SAP62, SF3a66
Chromosome Location
Chr. 19: 2236817 - 2248679 on Build GRCh38
UniGene ID
Hs.115232
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_007165.4 NP_009096.2 2, 3 256
GenBank mRNA AK314815.1 - 2, 3 249
BC004434.1 - 2, 3 226 2820900
BC009903.2 - 2, 3 249 2959124
BC015804.1 - 2, 3 209
BG325385.1 - 548
BQ642367.1 - 329

View Full Product Details