Gene Transcripts

Gene Symbol
Lgals3bp
Entrez Gene ID
245955
Gene Name
lectin, galactoside-binding, soluble, 3 binding protein
Gene Aliases
CyCAP, MAC2BP, Ppicap, mama
Chromosome Location
Chr. 10: 107415334 - 107424672 on Build Rnor_6.0
UniGene ID
Rn.3251
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139096.1 NP_620796.1 6 817
GenBank mRNA AF065438.1 - 6 817
AY552591.1 - 5 679
BC081724.1 - 6 790 7123606

View Full Product Details