Gene Transcripts
- Gene Symbol
- Clrn1
- Entrez Gene ID
- 261738
- Gene Name
- clarin 1
- Gene Aliases
- Ush3, Ush3a
- Chromosome Location
- Chr. 2: 149049925 - 149088787 on Build Rnor_6.0
- UniGene ID
- Rn.57072
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF482698.1 |
- |
1 |
143 |
|
View Full Product Details