Gene Transcripts
- Gene Symbol
- Cyp2b12
- Entrez Gene ID
- 29295
- Gene Name
- cytochrome P450, family 2, subfamily b, polypeptide 12
- Gene Aliases
- Cyp2b15
- Chromosome Location
- Chr. 1: 83511167 - 83522965 on Build Rnor_6.0
- UniGene ID
- Rn.144570
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017156.1 |
NP_058852.1
|
|
442 |
|
GenBank mRNA |
X63545.1 |
- |
|
442 |
|
View Full Product Details