Gene Transcripts

Gene Symbol
Nop58
Entrez Gene ID
60373
Gene Name
NOP58 ribonucleoprotein
Gene Aliases
Nap65, Nol5
Chromosome Location
Chr. 9: 66495881 - 66519790 on Build Rnor_6.0
UniGene ID
Rn.43930
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021754.1 NP_068522.1 3, 4 337
GenBank mRNA AF194371.1 - 3, 4 337
BC087637.1 - 3, 4 370 7309062

View Full Product Details