Gene Transcripts

Gene Symbol
B3gat2
Entrez Gene ID
64544
Gene Name
beta-1,3-glucuronyltransferase 2
Gene Aliases
Chromosome Location
Chr. 9: 29984077 - 30068649 on Build Rnor_6.0
UniGene ID
Rn.42869
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022609.2 NP_072131.1 1 759
GenBank mRNA AB010441.1 - 1 759
AF106624.1 - 1 754

View Full Product Details