Gene Transcripts

Gene Symbol
Cd40lg
Entrez Gene ID
84349
Gene Name
CD40 ligand
Gene Aliases
Cd40l, Tnfsf5
Chromosome Location
Chr. X: 159703703 - 159714886 on Build Rnor_6.0
UniGene ID
Rn.44218
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053353.1 NP_445805.1 2 188
GenBank mRNA AF013985.1 - 2 188
AF116582.1 - 2 188
EF066490.1 - 2 188

View Full Product Details