Gene Transcripts

Gene Symbol
Tmf1
Entrez Gene ID
114206
Gene Name
TATA element modulatory factor 1
Gene Aliases
Chromosome Location
Chr. 4: 129521986 - 129546028 on Build Rnor_6.0
UniGene ID
Rn.91637
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053671.2 NP_446123.2 1, 2 223
XM_008763143.1 XP_008761365.1 1, 2 1745
GenBank mRNA AF107843.1 - 1, 2 146
BF415257.1 - 1, 2 223

View Full Product Details