Gene Transcripts

Gene Symbol
F2rl1
Entrez Gene ID
116677
Gene Name
coagulation factor II (thrombin) receptor-like 1
Gene Aliases
Par-2, Par2
Chromosome Location
Chr. 2: 25222324 - 25235275 on Build Rnor_6.0
UniGene ID
Rn.10543
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053897.2 NP_446349.1 1, 2 186
GenBank mRNA BC099765.1 - 1, 2 186
U61373.1 - 1, 2 99

View Full Product Details