Gene Transcripts

Gene Symbol
Rasd2
Entrez Gene ID
171099
Gene Name
RASD family, member 2
Gene Aliases
Rhes
Chromosome Location
Chr. 19: 14653198 - 14663914 on Build Rnor_6.0
UniGene ID
Rn.115571
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133568.1 NP_598252.1 2 1080
GenBank mRNA AF134409.1 - 2 1080

View Full Product Details