Gene Transcripts

Gene Symbol
Slc12a5
Entrez Gene ID
171373
Gene Name
solute carrier family 12 (potassium-chloride transporter), member 5
Gene Aliases
Kcc2
Chromosome Location
Chr. 3: 161433303 - 161465078 on Build Rnor_6.0
UniGene ID
Rn.10513
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134363.1 NP_599190.1 7 793
GenBank mRNA EF641113.1 - 806
U55816.1 - 7 793

View Full Product Details