Gene Transcripts

Gene Symbol
Ipmk
Entrez Gene ID
171458
Gene Name
inositol polyphosphate multikinase
Gene Aliases
Impk, Ipk2
Chromosome Location
Chr. 20: 18457309 - 18489890 on Build Rnor_6.0
UniGene ID
Rn.15187
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134417.1 NP_599244.1 4 488
XM_006256345.2 XP_006256407.1 4 544
XM_006256346.2 XP_006256408.1 4 906
XM_008772858.1 XP_008771080.1 4 420
GenBank mRNA AY014898.1 - 4 488

View Full Product Details