Gene Transcripts
- Gene Symbol
- Akr1c14
- Entrez Gene ID
- 191574
- Gene Name
- aldo-keto reductase family 1, member C14
- Gene Aliases
- Akr1c9
- Chromosome Location
- Chr. 17: 69806095 - 69827110 on Build Rnor_6.0
- UniGene ID
- Rn.10021
- Species
- Rattus norvegicus
- Gene Symbol
- Akr1c19
- Entrez Gene ID
- 307096
- Gene Name
- aldo-keto reductase family 1, member C19
- Gene Aliases
- RGD1562954
- Chromosome Location
- Chr. 17: 69653368 - 69675204 on Build Rnor_6.0
- UniGene ID
- Rn.206655
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
S35752.1 |
- |
|
74 |
|
View Full Product Details