Gene Transcripts

Gene Symbol
Exoc8
Entrez Gene ID
245709
Gene Name
exocyst complex component 8
Gene Aliases
Exo84
Chromosome Location
Chr. 19: 57647305 - 57649794 on Build Rnor_6.0
UniGene ID
Rn.10901
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139043.1 NP_620612.1 1 671
GenBank mRNA AF032669.1 - 1 671

View Full Product Details