Gene Transcripts

Gene Symbol
Rwdd1
Entrez Gene ID
259218
Gene Name
RWD domain containing 1
Gene Aliases
sarip
Chromosome Location
Chr. 20: 27535316 - 27552225 on Build Rnor_6.0
UniGene ID
Rn.234293
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_147146.2 NP_671487.1 7 980
XM_008772869.1 XP_008771091.1 7 1112
GenBank mRNA FQ218614.1 - 4 515
FQ220470.1 - 4 515

View Full Product Details