Gene Transcripts

Gene Symbol
Isca1
Entrez Gene ID
290985
Gene Name
iron-sulfur cluster assembly 1
Gene Aliases
Hbld2, hIscA
Chromosome Location
Chr. 17: 5281727 - 5294386 on Build Rnor_6.0
UniGene ID
Rn.1180
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181626.3 NP_853657.1 4 577
GenBank mRNA BC070929.1 - 4 577
BC078677.1 - 4 587 7133788

View Full Product Details