Gene Transcripts

Gene Symbol
Luzp1
Entrez Gene ID
79428
Gene Name
leucine zipper protein 1
Gene Aliases
Luzp
Chromosome Location
Chr. 5: 154831702 - 154907646 on Build Rnor_6.0
UniGene ID
Rn.154410
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030830.2 NP_110457.2 4 486
XM_006239274.2 XP_006239336.1 3 429
GenBank mRNA AF181259.1 - 5 2436

View Full Product Details