Gene Transcripts

Gene Symbol
Slc26a5
Entrez Gene ID
83819
Gene Name
solute carrier family 26 (anion exchanger), member 5
Gene Aliases
Pres, prestin
Chromosome Location
Chr. 4: 9821541 - 9860571 on Build Rnor_6.0
UniGene ID
Rn.64631
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030840.1 NP_110467.1 1, 2 149
GenBank mRNA AJ303372.1 - 1, 2 149

View Full Product Details