Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031704.2 | NP_113892.2 | 6 | 666 | |
XM_006230997.2 | XP_006231059.1 | 5 | 1209 | ||
GenBank mRNA | BC085724.1 | - | 5 | 992 | |
BC127489.1 | - | 6 | 666 | 7376571 | |
L20822.1 | - | 5 | 496 | ||
U87971.1 | - | 6 | 666 |