Gene Transcripts
- Gene Symbol
- OGFR
- Entrez Gene ID
- 11054
- Gene Name
- opioid growth factor receptor
- Gene Aliases
- Chromosome Location
- Chr. 20: 62804825 - 62814000 on Build GRCh38
- UniGene ID
- Hs.67896
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_007346.2 |
NP_031372.2
|
5, 6 |
502 |
|
GenBank mRNA |
AF109134.1 |
- |
5, 6 |
495 |
|
AF172449.1 |
- |
5, 6 |
500 |
|
AF172450.1 |
- |
5, 6 |
500 |
|
AF172451.1 |
- |
5, 6 |
500 |
|
AF172452.1 |
- |
5, 6 |
500 |
|
AF172453.1 |
- |
|
500 |
|
AK022234.1 |
- |
5, 6 |
492 |
|
AK024485.1 |
- |
3, 4 |
2588 |
|
AK314696.1 |
- |
5, 6 |
494 |
|
BC008768.2 |
- |
5, 6 |
477 |
3050950 |
BC014137.2 |
- |
5, 6 |
477 |
4643904 |
BC032666.1 |
- |
5, 6 |
457 |
5552531 |
BM551600.1 |
- |
5, 6 |
502 |
|
View Full Product Details