Gene Transcripts

Gene Symbol
Ctbp2
Entrez Gene ID
81717
Gene Name
C-terminal binding protein 2
Gene Aliases
Chromosome Location
Chr. 1: 205001354 - 205030565 on Build Rnor_6.0
UniGene ID
Rn.138124
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053335.1 NP_445787.1 1 473
GenBank mRNA AF222712.1 - 1 473

View Full Product Details