Gene Transcripts

Gene Symbol
Pip4k2c
Entrez Gene ID
140607
Gene Name
phosphatidylinositol-5-phosphate 4-kinase, type II, gamma
Gene Aliases
Pip5k2c
Chromosome Location
Chr. 7: 70483948 - 70498995 on Build Rnor_6.0
UniGene ID
Rn.94783
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080480.2 NP_536728.2 2 305
GenBank mRNA AF030558.1 - 2 305

View Full Product Details