Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_134331.1 | NP_599158.1 | 2 | 266 | |
XM_006237962.2 | XP_006238024.1 | 2 | 424 | ||
XM_006237963.2 | XP_006238025.1 | 2 | 424 | ||
XM_006237964.2 | XP_006238026.1 | 2 | 421 | ||
XM_006237965.2 | XP_006238027.1 | 2 | 422 | ||
XM_008763587.1 | XP_008761809.1 | 2 | 420 | ||
GenBank mRNA | U21954.1 | - | 2 | 266 | |
U21955.1 | - | 2 | 266 |