Gene Transcripts

Gene Symbol
Plod2
Entrez Gene ID
300901
Gene Name
procollagen lysine, 2-oxoglutarate 5-dioxygenase 2
Gene Aliases
Chromosome Location
Chr. 8: 99977334 - 100059736 on Build Rnor_6.0
UniGene ID
Rn.12945
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142915.1 NP_001136387.1 2 352
NM_175869.3 NP_787065.2 2 352
GenBank mRNA AJ430860.1 - 2 414
AJ430861.1 - 2 414

View Full Product Details