Gene Transcripts

Gene Symbol
Dfnb31
Entrez Gene ID
313255
Gene Name
deafness, autosomal recessive 31
Gene Aliases
Cip98, Whrn
Chromosome Location
Chr. 5: 79235541 - 79317206 on Build Rnor_6.0
UniGene ID
Rn.231801
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181088.1 NP_851602.1 1 587
GenBank mRNA AY227205.1 - 1 587

View Full Product Details